The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004729.3 | NP_004720.1 | 1 - 2 | 152 | 86 |
GenBank mRNA | AB018328.1 | - | 1 - 2 | 152 | 86 |