The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030928.3 | NP_112190.2 | 4 - 5 | 738 | 90 |
GenBank mRNA | AB053172.1 | - | 4 - 5 | 738 | 90 |
AF070552.1 | - | 2 - 3 | 279 | 90 | |
AF321125.1 | - | 4 - 5 | 709 | 90 | |
BC000137.2 | - | 4 - 5 | 700 | 90 | |
BC008676.1 | - | 4 - 5 | 576 | 90 | |
BC008860.2 | - | 4 - 5 | 631 | 90 | |
BC009410.1 | - | 4 - 5 | 694 | 90 | |
BC021126.1 | - | 2 - 3 | 270 | 90 | |
BC049205.1 | - | 2 - 3 | 342 | 90 |