The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004459.6 | NP_004450.3 | 5 - 6 | 2114 | 76 |
XM_005257150.3 | XP_005257207.1 | 5 - 6 | 2261 | 76 | |
XM_005257151.3 | XP_005257208.1 | 5 - 6 | 2261 | 76 | |
XM_005257152.2 | XP_005257209.1 | 5 - 6 | 2261 | 76 | |
XM_005257153.3 | XP_005257210.1 | 5 - 6 | 2261 | 76 | |
XM_005257154.3 | XP_005257211.1 | 5 - 6 | 2261 | 76 | |
XM_005257159.2 | XP_005257216.1 | 5 - 6 | 2261 | 76 | |
XM_005257160.2 | XP_005257217.1 | 5 - 6 | 2261 | 76 | |
XM_005257161.3 | XP_005257218.1 | 5 - 6 | 2261 | 76 | |
XM_017024353.1 | XP_016879842.1 | 5 - 6 | 2261 | 76 | |
GenBank mRNA | AK297281.1 | - | 6 - 7 | 1781 | 76 |
AY282495.1 | - | 5 - 6 | 1636 | 76 | |
U05237.1 | - | 5 - 6 | 1709 | 76 |