Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPTF
Entrez Gene ID
2186
Gene Name
bromodomain PHD finger transcription factor
Gene Aliases
FAC1, FALZ, NURF301
Chromosome Location
Chr.17: 67825517 - 67984378 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004459.6 NP_004450.3 21 - 22 6916 90
NM_182641.3 NP_872579.2 19 - 20 6538 90
XM_005257150.3 XP_005257207.1 22 - 23 7225 90
XM_005257151.3 XP_005257208.1 22 - 23 7222 90
XM_005257152.2 XP_005257209.1 22 - 23 7225 90
XM_005257153.3 XP_005257210.1 21 - 22 7063 90
XM_005257154.3 XP_005257211.1 22 - 23 7225 90
XM_005257155.3 XP_005257212.1 21 - 22 7036 90
XM_005257156.3 XP_005257213.1 21 - 22 7036 90
XM_005257157.3 XP_005257214.1 21 - 22 7033 90
XM_005257158.3 XP_005257215.1 20 - 21 6847 90
XM_005257159.2 XP_005257216.1 22 - 23 7225 90
XM_005257160.2 XP_005257217.1 21 - 22 7063 90
XM_005257161.3 XP_005257218.1 22 - 23 7225 90
XM_011524520.2 XP_011522822.1 22 - 23 7222 90
XM_011524521.2 XP_011522823.1 21 - 22 7033 90
XM_011524522.2 XP_011522824.1 20 - 21 6844 90
XM_011524523.2 XP_011522825.1 19 - 20 6682 90
XM_011524524.2 XP_011522826.1 14 - 15 4186 90
XM_011524525.2 XP_011522827.1 19 - 20 6682 90
XM_011524526.2 XP_011522828.1 19 - 20 6682 90
XM_017024353.1 XP_016879842.1 22 - 23 7225 90
XM_017024354.1 XP_016879843.1 16 - 17 5954 90
GenBank mRNA AB032251.1 - 19 - 20 6531 90
AK022570.1 - 7 - 8 1016 90
AK027184.1 - 1 - 2 92 90
AY282495.1 - 21 - 22 6438 90

View Full Product Details