The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145212.3 | NP_660213.1 | 4 - 5 | 465 | 146 |
NR_028356.1 | 4 - 5 | 465 | 146 | ||
GenBank mRNA | AK225894.1 | - | 4 - 5 | 361 | 146 |
BC000217.1 | - | 4 - 5 | 355 | 146 | |
BC022391.1 | - | 4 - 5 | 429 | 146 | |
BG532984.1 | - | 4 - 5 | 347 | 146 | |
BU154006.1 | - | - | 338 | 146 | |
BU533068.1 | - | 4 - 5 | 395 | 146 | |
DA310406.1 | - | 4 - 5 | 466 | 146 |