The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001815.4 | NP_001806.2 | 3 - 4 | 631 | 65 |
NR_102333.2 | 4 - 5 | 1135 | 65 | ||
GenBank mRNA | BC106728.1 | - | 3 - 4 | 569 | 65 |
D90277.1 | - | 3 - 4 | 617 | 65 | |
D90278.1 | - | 4 - 5 | 1108 | 65 | |
E03348.1 | - | 3 - 4 | 617 | 65 | |
E03349.1 | - | 5 - 6 | 990 | 65 | |
L00692.1 | - | 3 - 4 | 597 | 65 |