The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080744.1 | NP_001074213.1 | 5 - 6 | 908 | 74 |
NM_001080745.1 | NP_001074214.1 | 5 - 6 | 908 | 74 | |
NM_001346.2 | NP_001337.2 | 5 - 6 | 908 | 74 | |
GenBank mRNA | AB209033.1 | - | 5 - 6 | 751 | 74 |
AK304440.1 | - | 5 - 6 | 906 | 74 | |
AK314192.1 | - | 5 - 6 | 853 | 74 | |
AY737485.1 | - | 4 - 5 | 603 | 74 | |
BC089411.1 | - | 5 - 6 | 662 | 74 | |
BC112363.1 | - | 4 - 5 | 631 | 74 | |
D26135.1 | - | 5 - 6 | 887 | 74 |