The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006219.2 | NP_006210.1 | 1 - 2 | 189 | 76 |
XM_005247530.2 | XP_005247587.1 | 3 - 4 | 538 | 76 | |
XM_006713659.3 | XP_006713722.1 | 2 - 3 | 409 | 76 | |
XM_011512895.2 | XP_011511197.1 | 4 - 5 | 534 | 76 | |
XM_017006619.1 | XP_016862108.1 | 2 - 3 | 212 | 76 | |
GenBank mRNA | BC114432.1 | - | 1 - 2 | 177 | 76 |
S67334.1 | - | 1 - 2 | 173 | 76 |