Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNM3
Entrez Gene ID
26052
Gene Name
dynamin 3
Gene Aliases
Dyna III
Chromosome Location
Chr.1: 171841478 - 172418466 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136127.2 NP_001129599.1 13 - 14 1723 82
NM_001278252.1 NP_001265181.1 13 - 14 1723 82
NM_015569.4 NP_056384.2 13 - 14 1723 82
XM_017000978.1 XP_016856467.1 13 - 14 1706 82
XM_017000983.1 XP_016856472.1 13 - 14 1706 82
XM_017000984.1 XP_016856473.1 13 - 14 1706 82
XM_017000985.1 XP_016856474.1 13 - 14 1706 82
GenBank mRNA AB020627.2 - 13 - 14 1674 82
AK308992.1 - 13 - 14 1672 82
AL136712.1 - 13 - 14 1701 82
BC064546.1 - 13 - 14 1564 82

View Full Product Details