Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNM3
Entrez Gene ID
26052
Gene Name
dynamin 3
Gene Aliases
Dyna III
Chromosome Location
Chr.1: 171841478 - 172418466 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136127.2 NP_001129599.1 4 - 5 770 78
NM_001278252.1 NP_001265181.1 4 - 5 770 78
NM_015569.4 NP_056384.2 4 - 5 770 78
XM_005245079.1 XP_005245136.1 4 - 5 753 78
XM_005245080.1 XP_005245137.1 4 - 5 753 78
XM_006711268.2 XP_006711331.1 4 - 5 753 78
XM_017000976.1 XP_016856465.1 4 - 5 753 78
XM_017000977.1 XP_016856466.1 4 - 5 753 78
XM_017000978.1 XP_016856467.1 4 - 5 753 78
XM_017000979.1 XP_016856468.1 4 - 5 753 78
XM_017000980.1 XP_016856469.1 4 - 5 471 78
XM_017000981.1 XP_016856470.1 4 - 5 753 78
XM_017000982.1 XP_016856471.1 4 - 5 753 78
XM_017000983.1 XP_016856472.1 4 - 5 753 78
XM_017000984.1 XP_016856473.1 4 - 5 753 78
XM_017000985.1 XP_016856474.1 4 - 5 753 78
XM_017000986.1 XP_016856475.1 4 - 5 1094 78
XM_017000987.1 XP_016856476.1 4 - 5 625 78
XM_017000988.1 XP_016856477.1 4 - 5 753 78
XM_017000989.1 XP_016856478.1 4 - 5 753 78
XM_017000990.1 XP_016856479.1 4 - 5 753 78
XM_017000991.1 XP_016856480.1 4 - 5 753 78
XM_017000992.1 XP_016856481.1 4 - 5 753 78
GenBank mRNA AB020627.2 - 4 - 5 721 78
AK056279.1 - 4 - 5 750 78
AK307806.1 - 4 - 5 708 78
AK308992.1 - 4 - 5 719 78
AL136712.1 - 4 - 5 748 78
BC064546.1 - 4 - 5 611 78

View Full Product Details