The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006529.3 | NP_006520.2 | 8 - 9 | 1574 | 123 |
XM_011532268.2 | XP_011530570.1 | 4 - 5 | 820 | 123 | |
XM_017008630.1 | XP_016864119.1 | 9 - 10 | 1598 | 123 | |
GenBank mRNA | AF018157.1 | - | 2 - 3 | 337 | 123 |