The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127698.1 | NP_001121170.1 | 20 - 21 | 1961 | 106 |
NM_001127699.1 | NP_001121171.1 | 20 - 21 | 1961 | 106 | |
NM_006846.3 | NP_006837.2 | 20 - 21 | 1961 | 106 | |
XM_011537551.2 | XP_011535853.1 | 20 - 21 | 2047 | 106 | |
GenBank mRNA | AJ228139.2 | - | 20 - 21 | 1931 | 106 |
AK299455.1 | - | 20 - 21 | 1896 | 106 | |
AK301660.1 | - | 20 - 21 | 1894 | 106 | |
AK307588.1 | - | - | 1896 | 106 | |
AK307648.1 | - | 20 - 21 | 2052 | 106 | |
DQ149927.1 | - | 20 - 21 | 1961 | 106 | |
DQ149928.1 | - | 20 - 21 | 1961 | 106 | |
DQ149929.1 | - | 20 - 21 | 1961 | 106 |