The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127698.1 | NP_001121170.1 | 32 - 33 | 3259 | 65 |
NM_006846.3 | NP_006837.2 | 31 - 32 | 3169 | 65 | |
XM_011537551.2 | XP_011535853.1 | 32 - 33 | 3345 | 65 | |
GenBank mRNA | AF086524.1 | - | 3 - 4 | 326 | 65 |
AI341060.1 | - | 2 - 3 | 65 | ||
AJ228139.2 | - | 31 - 32 | 3139 | 65 | |
DQ149927.1 | - | 31 - 32 | 3169 | 65 | |
DQ149928.1 | - | 32 - 33 | 3259 | 65 | |
DR421881.1 | - | 3 - 4 | 284 | 65 |