Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPINK5
Entrez Gene ID
11005
Gene Name
serine peptidase inhibitor, Kazal type 5
Gene Aliases
LEKTI, LETKI, NETS, NS, VAKTI
Chromosome Location
Chr.5: 148063972 - 148137362 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127698.1 NP_001121170.1 5 - 6 485 102
NM_001127699.1 NP_001121171.1 5 - 6 485 102
NM_006846.3 NP_006837.2 5 - 6 485 102
XM_011537551.2 XP_011535853.1 5 - 6 571 102
GenBank mRNA AJ228139.2 - 5 - 6 455 102
AK299455.1 - 5 - 6 420 102
AK301660.1 - 5 - 6 418 102
AK307648.1 - 5 - 6 576 102
AK309753.1 - 5 - 6 477 102
DQ149927.1 - 5 - 6 485 102
DQ149928.1 - 5 - 6 485 102
DQ149929.1 - 5 - 6 485 102

View Full Product Details