Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCYL3
Entrez Gene ID
57147
Gene Name
SCY1 like pseudokinase 3
Gene Aliases
PACE-1, PACE1
Chromosome Location
Chr.1: 169853074 - 169894267 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020423.6 NP_065156.5 8 - 9 1043 146
NM_181093.3 NP_851607.2 8 - 9 1043 146
XM_006711465.1 XP_006711528.1 8 - 9 992 146
XM_011509801.1 XP_011508103.1 8 - 9 1132 146
XM_011509802.1 XP_011508104.1 8 - 9 992 146
XM_011509803.1 XP_011508105.1 7 - 8 895 146
XM_017001862.1 XP_016857351.1 8 - 9 992 146
XM_017001863.1 XP_016857352.1 8 - 9 1132 146
XM_017001864.1 XP_016857353.1 8 - 9 992 146
XM_017001865.1 XP_016857354.1 8 - 9 1091 146
GenBank mRNA AF540957.1 - 7 - 8 873 146
AK292507.1 - 8 - 9 989 146
AK304481.1 - 3 - 4 635 146
AL117233.1 - 8 - 9 926 146
AY144493.1 - 7 - 8 873 146
BC014662.1 - 8 - 9 980 146
BX647352.1 - 8 - 9 1132 146

View Full Product Details