The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138691.2 | NP_619636.2 | 23 - 24 | 2800 | 137 |
XM_017014256.1 | XP_016869745.1 | 20 - 21 | 2523 | 137 | |
GenBank mRNA | AF417578.1 | - | 23 - 24 | 2800 | 137 |
AK098607.1 | - | 10 - 11 | 1158 | 137 | |
BC103955.1 | - | 19 - 20 | 2366 | 137 | |
BC103956.1 | - | 18 - 19 | 2162 | 137 |