The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014319.4 | NP_055134.2 | 2 - 3 | 1586 | 99 |
GenBank mRNA | AF112299.2 | - | 2 - 3 | 1566 | 99 |