Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LEMD3
Entrez Gene ID
23592
Gene Name
LEM domain containing 3
Gene Aliases
MAN1
Chromosome Location
Chr.12: 65169571 - 65248361 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167614.1 NP_001161086.1 5 - 6 1798 133
NM_014319.4 NP_055134.2 5 - 6 1801 133
GenBank mRNA AF112299.2 - 5 - 6 1781 133
AK024954.1 - 5 - 6 476 133
AK312473.1 - 5 - 6 1798 133

View Full Product Details