The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167614.1 | NP_001161086.1 | 9 - 10 | 2324 | 152 |
NM_014319.4 | NP_055134.2 | 9 - 10 | 2327 | 152 | |
GenBank mRNA | AF112299.2 | - | 9 - 10 | 2307 | 152 |
AK024954.1 | - | 9 - 10 | 1002 | 152 | |
AK312473.1 | - | 9 - 10 | 2324 | 152 | |
AL137533.1 | - | 1 - 2 | 163 | 152 |