The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307990.1 | NP_001294919.1 | 26 - 27 | 1997 | 78 |
NM_004579.4 | NP_004570.2 | 26 - 27 | 2021 | 78 | |
XM_011545203.2 | XP_011543505.1 | 26 - 27 | 2015 | 78 | |
XM_011545204.2 | XP_011543506.1 | 25 - 26 | 1883 | 78 | |
XM_017018093.1 | XP_016873582.1 | 25 - 26 | 1859 | 78 | |
XM_017018094.1 | XP_016873583.1 | 25 - 26 | 1915 | 78 | |
XM_017018095.1 | XP_016873584.1 | 25 - 26 | 1915 | 78 | |
GenBank mRNA | BC039839.1 | - | 26 - 27 | 1904 | 78 |
BC047865.1 | - | 26 - 27 | 1984 | 78 | |
U07349.1 | - | 26 - 27 | 1952 | 78 |