Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL4
Entrez Gene ID
3565
Gene Name
interleukin 4
Gene Aliases
BCGF-1, BCGF1, BSF-1, BSF1, IL-4
Chromosome Location
Chr.5: 132673986 - 132682678 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000589.3 NP_000580.1 2 - 3 248 70
GenBank mRNA AM937235.1 - 2 - 3 223 70
BC066277.1 - 2 - 3 247 70
BC066278.1 - 2 - 3 248 70
BC067514.1 - 2 - 3 248 70
BC067515.1 - 2 - 3 248 70
BC070123.1 - 2 - 3 245 70
CD640452.1 - 2 - 3 248 70
EF173851.1 - 1 - 2 53 70
M13982.1 - 2 - 3 246 70

View Full Product Details