The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130777.2 | NP_570133.1 | 1 - 2 | 196 | 97 |
GenBank mRNA | AJ318880.1 | - | 1 - 2 | 171 | 97 |
BC009232.2 | - | 1 - 2 | 176 | 97 | |
BC066311.1 | - | 1 - 2 | 178 | 97 | |
H53211.1 | - | 1 - 2 | 196 | 97 |