The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152649.3 | NP_689862.1 | 6 - 7 | 1655 | 78 |
XM_005255834.3 | XP_005255891.1 | 7 - 8 | 1554 | 78 | |
GenBank mRNA | AK091708.1 | - | 6 - 7 | 1398 | 78 |
AL833362.1 | - | 5 - 6 | 1950 | 78 |