Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF2
Entrez Gene ID
7549
Gene Name
zinc finger protein 2
Gene Aliases
A1-5, ZNF661, Zfp661
Chromosome Location
Chr.2: 95165417 - 95184317 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282398.1 NP_001269327.1 1 - 2 447 90
NM_001291605.1 NP_001278534.1 1 - 2 447 90
NM_021088.3 NP_066574.2 1 - 2 447 90
GenBank mRNA AK055072.1 - 1 - 2 426 90
AK295739.1 - 1 - 2 75 90
BC005068.1 - 1 - 2 419 90

View Full Product Details