Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM63A
Entrez Gene ID
55793
Gene Name
family with sequence similarity 63 member A
Gene Aliases
-
Chromosome Location
Chr.1: 150996737 - 151008378 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040217.2 NP_001035307.1 9 - 10 1633 148
NM_001163258.1 NP_001156730.1 10 - 11 1696 148
NM_001163259.1 NP_001156731.1 9 - 10 1598 148
NM_001163260.1 NP_001156732.1 9 - 10 1509 148
NM_001319998.1 NP_001306927.1 10 - 11 3013 148
NM_018379.4 NP_060849.2 10 - 11 2297 148
XM_005245321.3 XP_005245378.1 9 - 10 2274 148
XM_005245325.3 XP_005245382.1 9 - 10 1629 148
XM_005245327.3 XP_005245384.1 8 - 9 1470 148
XM_005245328.3 XP_005245385.1 8 - 9 1467 148
XM_011509757.2 XP_011508059.1 9 - 10 2247 148
XM_011509759.2 XP_011508061.1 8 - 9 1850 148
XM_017001772.1 XP_016857261.1 9 - 10 2009 148
XM_017001773.1 XP_016857262.1 9 - 10 2244 148
XM_017001774.1 XP_016857263.1 9 - 10 1982 148
XM_017001775.1 XP_016857264.1 8 - 9 2112 148
XM_017001776.1 XP_016857265.1 8 - 9 2085 148
XM_017001777.1 XP_016857266.1 9 - 10 1623 148
GenBank mRNA AB037811.1 - 8 - 9 1482 148
AK002142.1 - 9 - 10 1359 148
AK125493.1 - 9 - 10 1333 148
AK125959.1 - 10 - 11 3013 148
AK301946.1 - 10 - 11 1696 148
AK303962.1 - 9 - 10 1286 148
BC032321.1 - 9 - 10 1773 148
BC075812.1 - 10 - 11 2284 148

View Full Product Details