Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SVIL
Entrez Gene ID
6840
Gene Name
supervillin
Gene Aliases
-
Chromosome Location
Chr.10: 29457338 - 29737001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323599.1 NP_001310528.1 25 - 26 4273 83
NM_001323600.1 NP_001310529.1 23 - 24 3874 83
NM_003174.3 NP_003165.2 22 - 23 3659 83
NM_021738.2 NP_068506.2 24 - 25 5240 83
XM_005252570.2 XP_005252627.1 28 - 29 5263 83
XM_005252571.3 XP_005252628.1 28 - 29 5189 83
XM_005252573.2 XP_005252630.1 26 - 27 5395 83
XM_011519630.2 XP_011517932.2 25 - 26 6030 83
XM_011519631.2 XP_011517933.2 25 - 26 6012 83
XM_011519632.2 XP_011517934.2 25 - 26 5937 83
XM_011519633.2 XP_011517935.2 23 - 24 5886 83
XM_011519634.1 XP_011517936.1 28 - 29 5321 83
XM_011519635.1 XP_011517937.1 26 - 27 5288 83
XM_011519636.2 XP_011517938.2 25 - 26 5178 83
XM_011519637.2 XP_011517939.2 23 - 24 4926 83
XM_011519638.2 XP_011517940.2 22 - 23 4830 83
XM_017016575.1 XP_016872064.1 26 - 27 6108 83
XM_017016576.1 XP_016872065.1 25 - 26 6027 83
XM_017016577.1 XP_016872066.1 24 - 25 5982 83
XM_017016578.1 XP_016872067.1 24 - 25 5082 83
XM_017016579.1 XP_016872068.1 23 - 24 5052 83
XM_017016580.1 XP_016872069.1 24 - 25 5007 83
XM_017016581.1 XP_016872070.1 21 - 22 4800 83
XM_017016582.1 XP_016872071.1 20 - 21 4704 83
GenBank mRNA AF051850.1 - 22 - 23 3659 83
AF051851.1 - 18 - 19 3309 83
AF109135.1 - 24 - 25 5240 83
AK302077.1 - 7 - 8 945 83
AK304720.1 - 10 - 11 1265 83
AK304799.1 - 8 - 9 1023 83
AK310272.1 - 7 - 8 906 83
AL117410.1 - 9 - 10 1317 83
JX467682.1 - 19 - 20 4391 83

View Full Product Details