Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ANKRD26P3
- Entrez Gene ID
-
100101938
- Gene Name
- ankyrin repeat domain 26 pseudogene 3
- Gene Aliases
-
LINC00414
- Chromosome Location
-
Chr.13: 19262800 - 19344973 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027248.3 |
|
12 - 13 |
1806 |
72 |
GenBank mRNA |
BC036210.1 |
- |
12 - 13 |
1806 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101928697 |
LOC101928697 |
|
-107.1kb
|
Anti-sense |
Chr. - 19182033 - 19187465 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 421 |
LINC00421 |
|
49.7kb
|
Anti-sense |
Chr. - 19345049 - 19346749 on Build GRCh38 |
View Full Product Details