The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048200.2 | NP_001041665.1 | 5 - 6 | 2001 | 114 |
NM_001278162.1 | NP_001265091.1 | 5 - 6 | 1793 | 114 | |
NM_001278163.1 | NP_001265092.1 | 5 - 6 | 1514 | 114 | |
NM_005734.4 | NP_005725.3 | 5 - 6 | 2001 | 114 | |
XM_005252729.2 | XP_005252786.1 | 5 - 6 | 1788 | 114 | |
XM_017017076.1 | XP_016872565.1 | 4 - 5 | 1435 | 114 | |
XM_017017077.1 | XP_016872566.1 | 5 - 6 | 1458 | 114 | |
GenBank mRNA | AF004849.1 | - | 4 - 5 | 1440 | 114 |
AF305239.1 | - | 5 - 6 | 1464 | 114 | |
AK293009.1 | - | 5 - 6 | 1794 | 114 | |
AK315277.1 | - | 5 - 6 | 1514 | 114 |