Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CIITA
- Entrez Gene ID
-
4261
- Gene Name
- class II, major histocompatibility complex, transactivator
- Gene Aliases
-
C2TA, CIITAIV, MHC2TA, NLRA
- Chromosome Location
-
Chr.16: 10866208 - 10941562 on Build GRCh38
View Full Product Details