The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258384.2 | NP_001245313.1 | 1 - 2 | 68 | 109 |
NM_006899.4 | NP_008830.2 | 1 - 2 | 68 | 109 | |
NM_174855.3 | NP_777280.1 | 1 - 2 | 68 | 109 | |
NR_136344.1 | 1 - 2 | 68 | 109 | ||
XM_005260716.1 | XP_005260773.1 | 1 - 2 | 61 | 109 | |
GenBank mRNA | AK300485.1 | - | 1 - 2 | 54 | 109 |
AK315641.1 | - | 1 - 2 | 50 | 109 | |
BX647393.1 | - | 1 - 2 | 86 | 109 | |
HY131986.1 | - | 1 - 2 | 69 | 109 |