Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR3B
Entrez Gene ID
55703
Gene Name
RNA polymerase III subunit B
Gene Aliases
C128, HLD8, INMAP, RPC2
Chromosome Location
Chr.12: 106357658 - 106510198 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160708.1 NP_001154180.1 14 - 15 1534 97
NM_018082.5 NP_060552.4 14 - 15 1687 97
XM_017019621.1 XP_016875110.1 14 - 15 1652 97
GenBank mRNA AK092031.1 - 4 - 5 610 97
AK122713.1 - 14 - 15 1514 97
AK291635.1 - 14 - 15 1531 97
AY092084.1 - 14 - 15 1518 97
BC046238.1 - 14 - 15 1554 97
BX537447.1 - 14 - 15 1635 97

View Full Product Details