The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166621.1 | NP_001160093.1 | 1 - 2 | 63 | 76 |
NR_030697.1 | 1 - 2 | 63 | 76 | ||
GenBank mRNA | BI600633.1 | - | 1 - 2 | 68 | 76 |