Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLTM
Entrez Gene ID
79811
Gene Name
SAFB like transcription modulator
Gene Aliases
Met
Chromosome Location
Chr.15: 58879045 - 58934313 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013843.2 NP_001013865.1 10 - 11 1429 153
NM_024755.3 NP_079031.2 10 - 11 1483 153
NR_135042.1 8 - 9 816 153
NR_135043.1 8 - 9 926 153
XM_006720686.3 XP_006720749.3 8 - 9 1874 153
XM_006720690.1 XP_006720753.1 6 - 7 580 153
XM_011522022.1 XP_011520324.1 10 - 11 2141 153
XM_011522023.1 XP_011520325.1 10 - 11 2087 153
XM_011522025.1 XP_011520327.1 9 - 10 1943 153
XM_011522026.2 XP_011520328.2 9 - 10 1922 153
XM_011522027.1 XP_011520329.1 8 - 9 1895 153
XM_011522028.1 XP_011520330.1 8 - 9 1841 153
XM_011522029.2 XP_011520331.1 10 - 11 1322 153
XM_011522030.2 XP_011520332.1 10 - 11 2141 153
XM_011522031.2 XP_011520333.1 7 - 8 977 153
XM_017022577.1 XP_016878066.1 9 - 10 1868 153
XM_017022578.1 XP_016878067.1 8 - 9 1820 153
XM_017022579.1 XP_016878068.1 10 - 11 1486 153
XM_017022580.1 XP_016878069.1 9 - 10 940 153
GenBank mRNA AK000867.1 - 10 - 11 1375 153
AK024710.1 - 4 - 5 796 153
AK055195.1 - 8 - 9 719 153
AK291423.1 - 8 - 9 849 153
BC014944.1 - 3 - 4 286 153
BC140851.1 - 10 - 11 1450 153

View Full Product Details