The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024755.3 | NP_079031.2 | 5 - 6 | 670 | 92 |
XM_011522022.1 | XP_011520324.1 | 5 - 6 | 1328 | 92 | |
XM_011522024.1 | XP_011520326.1 | 5 - 6 | 1328 | 92 | |
XM_011522025.1 | XP_011520327.1 | 4 - 5 | 1130 | 92 | |
XM_011522026.2 | XP_011520328.2 | 4 - 5 | 1109 | 92 | |
XM_011522029.2 | XP_011520331.1 | 5 - 6 | 509 | 92 | |
XM_011522030.2 | XP_011520332.1 | 5 - 6 | 1328 | 92 | |
XM_017022579.1 | XP_016878068.1 | 5 - 6 | 673 | 92 | |
GenBank mRNA | AK000867.1 | - | 5 - 6 | 562 | 92 |
BC108656.1 | - | 5 - 6 | 594 | 92 | |
BC140851.1 | - | 5 - 6 | 637 | 92 |