The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271606.1 | NP_001258535.1 | 2 - 2 | 1537 | 117 |
NM_006317.4 | NP_006308.3 | 2 - 2 | 1712 | 117 | |
GenBank mRNA | AK295995.1 | - | 3 - 3 | 1479 | 118 |
BM888431.1 | - | 1 - 1 | 222 | 118 | |
BM973114.1 | - | 1 - 1 | 113 | ||
HY258599.1 | - | 1 - 1 | 117 |