The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301015.1 | NP_001287944.1 | 15 - 16 | 2103 | 94 |
NM_001301018.1 | NP_001287947.1 | 14 - 15 | 1478 | 94 | |
NM_001301027.1 | NP_001287956.1 | 12 - 13 | 1849 | 94 | |
NM_001301028.1 | NP_001287957.1 | 11 - 12 | 1770 | 94 | |
NM_014945.3 | NP_055760.1 | 16 - 17 | 1749 | 94 | |
XM_005268392.1 | XP_005268449.1 | 15 - 16 | 1631 | 94 | |
XM_005268393.1 | XP_005268450.1 | 14 - 15 | 1583 | 94 | |
XM_005268394.1 | XP_005268451.1 | 14 - 15 | 1544 | 94 | |
XM_005268395.1 | XP_005268452.1 | 13 - 14 | 1445 | 94 | |
XM_005268396.1 | XP_005268453.1 | 12 - 13 | 1397 | 94 | |
XM_017009238.1 | XP_016864727.1 | 12 - 13 | 1539 | 94 | |
GenBank mRNA | AB020650.1 | - | 16 - 17 | 1662 | 94 |
AB075881.1 | - | 1 - 2 | 1672 | 94 | |
AK225769.1 | - | 15 - 16 | 1971 | 94 | |
AK289736.1 | - | 15 - 16 | 1516 | 94 | |
AL833021.1 | - | 9 - 10 | 857 | 94 | |
BC001665.2 | - | 11 - 12 | 1432 | 94 | |
DQ413174.1 | - | 14 - 15 | 1440 | 94 |