The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282874.1 | NP_001269803.1 | 3 - 4 | 563 | 76 |
NM_001282875.1 | NP_001269804.1 | 3 - 4 | 563 | 76 | |
NM_003069.4 | NP_003060.2 | 3 - 4 | 563 | 76 | |
XM_005262461.2 | XP_005262518.1 | 3 - 4 | 578 | 76 | |
XM_005262462.2 | XP_005262519.1 | 3 - 4 | 581 | 76 | |
XM_006724782.2 | XP_006724845.1 | 3 - 4 | 622 | 76 | |
XM_017029750.1 | XP_016885239.1 | 3 - 4 | 622 | 76 | |
XM_017029751.1 | XP_016885240.1 | 3 - 4 | 622 | 76 | |
GenBank mRNA | AK026426.1 | - | 3 - 4 | 438 | 76 |
BC051825.1 | - | 2 - 3 | 240 | 76 | |
BC117447.1 | - | 3 - 4 | 463 | 76 | |
BC143954.1 | - | 3 - 4 | 463 | 76 | |
DR002115.1 | - | 3 - 4 | 567 | 76 | |
EU636009.1 | - | 3 - 4 | 522 | 76 | |
M89907.1 | - | 3 - 4 | 851 | 76 |