The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282874.1 | NP_001269803.1 | 9 - 10 | 1303 | 134 |
NM_001282875.1 | NP_001269804.1 | 9 - 10 | 1303 | 134 | |
NM_003069.4 | NP_003060.2 | 9 - 10 | 1303 | 134 | |
XM_005262461.2 | XP_005262518.1 | 9 - 10 | 1318 | 134 | |
XM_005262462.2 | XP_005262519.1 | 9 - 10 | 1321 | 134 | |
XM_006724782.2 | XP_006724845.1 | 9 - 10 | 1362 | 134 | |
XM_017029750.1 | XP_016885239.1 | 9 - 10 | 1362 | 134 | |
XM_017029751.1 | XP_016885240.1 | 9 - 10 | 1362 | 134 | |
GenBank mRNA | AK026426.1 | - | 9 - 10 | 1178 | 134 |
BC051825.1 | - | 8 - 9 | 980 | 134 | |
BC117447.1 | - | 9 - 10 | 1203 | 134 | |
BC143954.1 | - | 9 - 10 | 1203 | 134 | |
EU636009.1 | - | 9 - 10 | 1262 | 134 | |
M89907.1 | - | 9 - 10 | 1591 | 134 |