The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130831.1 | NP_001124303.1 | 10 - 11 | 990 | 60 |
NM_003644.2 | NP_003635.2 | 6 - 7 | 739 | 60 | |
NM_201432.1 | NP_958836.1 | 10 - 11 | 1147 | 60 | |
NM_201433.1 | NP_958839.1 | 10 - 11 | 1177 | 60 | |
XM_005256831.3 | XP_005256888.1 | 11 - 12 | 1088 | 60 | |
XM_005256832.3 | XP_005256889.1 | 10 - 11 | 958 | 60 | |
XM_005256833.3 | XP_005256890.1 | 9 - 10 | 1065 | 60 | |
XM_005256834.3 | XP_005256891.1 | 9 - 10 | 891 | 60 | |
XM_005256835.3 | XP_005256892.1 | 9 - 10 | 835 | 60 | |
XM_011524044.2 | XP_011522346.1 | 10 - 11 | 982 | 60 | |
XM_017025239.1 | XP_016880728.1 | 10 - 11 | 1011 | 60 | |
XM_017025240.1 | XP_016880729.1 | 9 - 10 | 794 | 60 | |
XM_017025241.1 | XP_016880730.1 | 9 - 10 | 770 | 60 | |
XM_017025242.1 | XP_016880731.1 | 9 - 10 | 944 | 60 | |
GenBank mRNA | AB007854.1 | - | 10 - 11 | 945 | 60 |
AB209030.1 | - | 9 - 10 | 834 | 60 | |
AJ224876.1 | - | 6 - 7 | 762 | 60 | |
AK057761.1 | - | 7 - 8 | 737 | 60 | |
AK225165.1 | - | 10 - 11 | 969 | 60 | |
AK292987.1 | - | 10 - 11 | 963 | 60 | |
AK293755.1 | - | 6 - 7 | 570 | 60 | |
AK293834.1 | - | 4 - 5 | 548 | 60 | |
AK294829.1 | - | 9 - 10 | 938 | 60 | |
AK307987.1 | - | 11 - 12 | 999 | 60 | |
AK309722.1 | - | 10 - 11 | 998 | 60 | |
AK315162.1 | - | 6 - 7 | 752 | 60 | |
AY327406.1 | - | 10 - 11 | 1177 | 60 | |
BC001152.1 | - | 10 - 11 | 970 | 60 | |
BC006454.1 | - | 10 - 11 | 1147 | 60 | |
BT006891.1 | - | 9 - 10 | 824 | 60 |