Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSRP3
Entrez Gene ID
8048
Gene Name
cysteine and glycine rich protein 3
Gene Aliases
CLP, CMD1M, CMH12, CRP3, LMO4, MLP
Chromosome Location
Chr.11: 19182030 - 19210571 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003476.4 NP_003467.1 4 - 5 515 56
GenBank mRNA AF121260.1 - 3 - 4 405 56
AK313289.1 - 3 - 4 340 56
BC005900.2 - 2 - 3 305 56
BC024010.1 - 4 - 5 518 56
BC057221.1 - 3 - 4 338 56
BF671742.1 - - 56
U20324.1 - 3 - 4 321 56
U49837.1 - 3 - 4 333 56
U72899.1 - 3 - 4 321 56

View Full Product Details