Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HORMAD1
Entrez Gene ID
84072
Gene Name
HORMA domain containing 1
Gene Aliases
CT46, NOHMA
Chromosome Location
Chr.1: 150698059 - 150720910 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199829.1 NP_001186758.1 10 - 11 960 153
NM_032132.4 NP_115508.2 11 - 12 981 153
XM_006711570.2 XP_006711633.1 10 - 11 875 153
XM_006711571.2 XP_006711634.1 7 - 8 737 153
XM_011510053.2 XP_011508355.1 11 - 12 1049 153
XM_011510054.2 XP_011508356.1 11 - 12 1003 153
XM_011510055.2 XP_011508357.1 10 - 11 799 153
XM_011510056.2 XP_011508358.1 6 - 7 581 153
GenBank mRNA AK097390.1 - 11 - 12 969 153
AK098633.1 - 11 - 12 939 153
AL136755.2 - 10 - 11 957 153
AY626344.1 - 10 - 11 957 153
BC047406.1 - 11 - 12 969 153
CR533505.1 - 9 - 10 842 153

View Full Product Details