Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A43
Entrez Gene ID
203427
Gene Name
solute carrier family 25 member 43
Gene Aliases
-
Chromosome Location
Chr.X: 119399295 - 119454478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145305.2 NP_660348.2 1 - 2 381 196
XM_017029340.1 XP_016884829.1 1 - 2 350 196
XM_017029341.1 XP_016884830.1 1 - 2 350 196
GenBank mRNA AK091304.1 - 1 - 2 296 196
AK292035.1 - 1 - 2 295 196
AK303932.1 - 1 - 2 296 196
BC019584.1 - 1 - 2 616 196
BC071871.1 - 1 - 2 343 196

View Full Product Details