The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004998.3 | NP_004989.2 | 10 - 11 | 1476 | 80 |
GenBank mRNA | AB290177.1 | - | 10 - 11 | 1459 | 80 |
AK291688.1 | - | 10 - 11 | 1473 | 80 | |
BC098392.1 | - | 10 - 11 | 1475 | 80 | |
U14391.1 | - | 10 - 11 | 1479 | 80 |