The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004998.3 | NP_004989.2 | 11 - 12 | 1556 | 125 |
GenBank mRNA | AB290177.1 | - | 11 - 12 | 1539 | 125 |
AK291688.1 | - | 11 - 12 | 1553 | 125 | |
BC098392.1 | - | 11 - 12 | 1555 | 125 | |
U14391.1 | - | 11 - 12 | 1559 | 125 |