The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083908.1 | NP_001077377.1 | 1 - 2 | 1948 | 91 |
GenBank mRNA | BC011400.1 | - | 1 - 2 | 712 | 91 |
BC013322.1 | - | 1 - 2 | 661 | 91 | |
BX248264.1 | - | 1 - 2 | 684 | 91 |