The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018077.2 | NP_060547.2 | 3 - 4 | 483 | 138 |
XM_011516370.2 | XP_011514672.1 | 4 - 5 | 590 | 138 | |
XM_017012389.1 | XP_016867878.1 | 4 - 5 | 591 | 138 | |
XM_017012390.1 | XP_016867879.1 | 3 - 4 | 483 | 138 | |
GenBank mRNA | AK001239.1 | - | 3 - 4 | 483 | 138 |
AK222716.1 | - | 3 - 4 | 445 | 138 | |
BC013889.2 | - | 3 - 4 | 437 | 138 | |
DA566130.1 | - | 3 - 4 | 483 | 138 | |
KU178685.1 | - | 3 - 4 | 368 | 138 |