Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THRSP
Entrez Gene ID
7069
Gene Name
thyroid hormone responsive
Gene Aliases
LPGP1, Lpgp, S14, SPOT14, THRP
Chromosome Location
Chr.11: 78063861 - 78068357 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003251.3 NP_003242.1 1 - 1 446 63
GenBank mRNA BC031989.1 - 1 - 1 429 63
BG674197.1 - 1 - 1 399 63

View Full Product Details