Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WAC
Entrez Gene ID
51322
Gene Name
WW domain containing adaptor with coiled-coil
Gene Aliases
BM-016, DESSH, PRO1741, Wwp4
Chromosome Location
Chr.10: 28532426 - 28623112 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL713727.1 - 7 - 8 1501 126

View Full Product Details