Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- WAC
- Entrez Gene ID
-
51322
- Gene Name
- WW domain containing adaptor with coiled-coil
- Gene Aliases
-
BM-016, DESSH, PRO1741, Wwp4
- Chromosome Location
-
Chr.10: 28532426 - 28623112 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF116666.2 |
- |
3 - 4 |
222 |
118 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BM712640.1 |
- |
1 - 2 |
77 |
|
Chr. 10 - 28534261 - 28589799 [+] on Build GRCh38 |
118 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
WAC antisense RNA 1 (head to head) |
WAC-AS1 |
|
-1.9kb
|
Anti-sense |
Chr. - 28519917 - 28532354 on Build GRCh38 |
Downstream |
BMP and activin membrane bound inhibitor |
BAMBI |
NMA |
141.7kb
|
Sense |
Chr. - 28677495 - 28682939 on Build GRCh38 |
View Full Product Details