The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048218.1 | NP_001041683.1 | 17 - 18 | 2332 | 67 |
NM_020680.3 | NP_065731.3 | 17 - 18 | 2383 | 67 | |
XM_005274118.3 | XP_005274175.1 | 17 - 18 | 2392 | 67 | |
XM_017018029.1 | XP_016873518.1 | 15 - 16 | 2568 | 67 | |
XM_017018030.1 | XP_016873519.1 | 17 - 18 | 2341 | 67 | |
XM_017018032.1 | XP_016873521.1 | 15 - 16 | 2410 | 67 | |
XM_017018033.1 | XP_016873522.1 | 17 - 18 | 2336 | 67 | |
GenBank mRNA | AB047077.1 | - | 16 - 17 | 2003 | 67 |
AB051427.1 | - | 17 - 18 | 2306 | 67 | |
AB051428.1 | - | 17 - 18 | 2255 | 67 | |
AF225424.1 | - | 11 - 12 | 1404 | 67 | |
AF297709.1 | - | 17 - 18 | 2305 | 67 | |
BC069233.1 | - | 17 - 18 | 2347 | 67 | |
BU728709.1 | - | 1 - 2 | 67 |